missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical™

Product Code. p-7073244 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 Tablets
500 Tablets
Unit Size:
1000 tablets
500 tablets
This item is not returnable. View return policy

Product Code. 10395850

Brand: Fisher Chemical K/0120/80

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kjeldahl Copper Catalyst Tablets, Technical, Contains 1g Na2SO4 And Equivalent Of 0.1g CuSO4, Fisher Chemical, Quantity: 1000 tablets, Blue-White, MDL Number: 147336, Packaging: Plastic powder jar, Physical Form: Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 147336

Specifications

Color Blue-White
Packaging Plastic powder jar
Quantity 500 Tablets
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.