missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KITLG, Mouse anti-Human, Clone: 2B1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004254-M09.100ug
This item is not returnable.
View return policy
Description
Sequence: EGICRNRVTNNVKDVTKLVANLPKDYMITLKYVPGMDVLPSHCWISEMVVQLSDSLTDLLDKFSNISEGLSNYSIIDKLVNIVDDLVECVKENSSKDLKKSFKSPEPRLFTPEEFFRIFNRSIDAFKDFVVASETSDCVVSSTLSPEKGKAKNPPGDSSLHWAAMALPALFSLIIGFAFGALYWKKRQPSLTRAVENIQINEEDNEISMLQEKEREFQEVSpecifications
| KITLG | |
| Monoclonal | |
| Unconjugated | |
| KIT ligand | |
| DKFZp686F2250/KL-1/Kitl/MGF/SCF/SF/SHEP7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2B1 | |
| In 1x PBS, pH 7.4 | |
| NM_003994 | |
| KITLG | |
| KITLG (NP_003985, 26 a.a. ∼ 245 a.a) partial recombinant protein. | |
| RUO | |
| 4254 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction