missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ KITLG (Human) Recombinant Protein

Product Code. p-7164816
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16242360

Brand: Abnova™ P5930.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human KITLG (181 a.a.) partial recombinant protein with His tag expressed in Barley grain (Hordeum vulgare). This protein is endotoxins-free and thus optimal to use in stem cell cultures.

Specifications

Gene ID (Entrez) 4254
Name KIT ligand
Preparation Method Hordeum vulgare (barley) expression system
Quantity 100 μg
Storage Requirements Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DKFZp686F2250, KL-1, Kitl, MGF, SCF, SF, SHEP7
Gene Symbol KITLG
Cross Reactivity Human
Species Plants
Protein Tag His
Buffer Lyophilized from PBS, pH 7.2
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.