missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIT [V654A] Recombinant Human Protein
KIT [V654A] Recombinant Human Protein
Brand: Invitrogen PV4132
Code : 50
Additional Details : Weight : 0.05250kg
Description
KIT is a proto-oncogene encoding a transmembrane tyrosine kinase receptor for stem cell factor required for normal hematopoiesis, melangenesis and gametogenesis. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. The clinically observed V654A mutant has been demonstrated to be resistant to inhibition with imatinib.Specifications
NP_000213, V654A | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
53.2kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
KIT V654A | |
KIT V654A was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 544-976; V654A | |
6.2nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 20μg/mL | |
His Tag | |
Kinase and inhibitors | |
50% by SDS-PAGE |
0.28mg/mL | |
3815, 4775 | |
KIT [V654A] Recombinant Human Protein | |
–80°C | |
1 Each | |
KIT [V654A] Recombinant Human Protein | |
PDGFR | |
KIT [V654A] Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.