missing translation for 'onlineSavingsMsg'
Learn More

KIST Antibody (OTI2H4), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18431025 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431025 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431025 Supplier Novus Biologicals Supplier No. NBP272383MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIST Monoclonal antibody specifically detects KIST in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIST
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2H4
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias DKFZp434C1613, EC 2.7.11.1, FLJ23015, kinase interacting with leukemia-associated gene (stathmin), Kinase interacting with stathmin, KISKIS protein kinase, KIST, serine/threonine-protein kinase Kist, U2AF homology motif (UHM) kinase 1, U2AF homology motif kinase 1
Host Species Mouse
Immunogen Full length human recombinant protein of human UHMK1 (NP_787062) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 127933
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.