missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KISS1R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527180
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GCPR54 is a galanin-like G protein coupled receptor that binds metastin, a peptide encoded by the metastasis suppressor gene KISS1. The tissue distribution of the protein suggests that it is involved in the regulation of endocrine function, and this is supported by the finding that it appears to play a role in the onset of puberty. GPR54 has been reported in various human brain regions, placenta, pancreas, and spinal cord, and at lower levels in spleen, peripheral blood leukocytes, testis, lymph node, thymus, small intestine, stomach, lung, and kidney. The receptor is also abundant in tumor tissues. In rat, GPR54 is expressed in liver, intestine, and in the pons, midbrain, thalamus, hypothalamus, hippocampus, amygdala, cortex (including frontal), and striatum. ESTs have been isolated from normal placenta and kidney cancer libraries.
Specifications
| KISS1R | |
| Polyclonal | |
| Unconjugated | |
| KISS1R | |
| AXOR12; CPPB1; G protein-coupled receptor 54; Gpr54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; HH8; HOT7T175; Hypogonadotropin-1; kiSS-; KiSS-1; KISS1 receptor; kiSS-1 receptor; KISS1R; KiSS-1R; Kisspeptins receptor; metastin receptor; mOT7T175; orphan G protein-coupled receptor 54; orphan G protein-coupled receptor GPR54; rOT7T175 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84634 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7 | |
| Q969F8 | |
| KISS1R | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of human GPR54. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction