missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KiSS1R/GPR54 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16724
This item is not returnable.
View return policy
Description
KiSS1R/GPR54 Polyclonal antibody specifically detects KiSS1R/GPR54 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| KiSS1R/GPR54 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 50% Glycerol with 0.01% Thimerosal | |
| AXOR12hypogonadotropin-1, G protein-coupled receptor 54, GPR54kiSS-1R, G-protein coupled receptor 54, G-protein coupled receptor OT7T175, HOT7T175, Hypogonadotropin-1, kiSS-1 receptor, KISS1 receptor, KiSS-1R, Kisspeptins receptor, Metastin receptor | |
| Rabbit | |
| 43 kDa | |
| 0.1 mL | |
| GPCR | |
| 84634 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q969F8 | |
| KISS1R | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GPR54. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction