missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KISS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557314
This item is not returnable.
View return policy
Description
Immunogen sequence: CTERKPAATA RLSRRGTSLS PPPESSGSPQ QPGLSAPHSR QIPAPQGAVL VQREKDLPNY NWNSFGLRFG KREAAPGNHG RS Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 51%.
This gene is a metastasis suppressor gene that suppresses metastases of melanomas and breast carcinomas without affecting tumorigenicity. The encoded protein may inhibit chemotaxis and invasion and thereby attenuate metastasis in malignant melanomas. Studies suggest a putative role in the regulation of events downstream of cell-matrix adhesion, perhaps involving cytoskeletal reorganization. A protein product of this gene, kisspeptin, stimulates gonadotropin-releasing hormone (GnRH)-induced gonadotropin secretion and regulates the pubertal activation of GnRH neurons. A polymorphism in the terminal exon of this mRNA results in two protein isoforms. An adenosine present at the polymorphic site represents the third position in a stop codon. When the adenosine is absent, a downstream stop codon is utilized and the encoded protein extends for an additional seven amino acid residues.
Specifications
| KISS1 | |
| Polyclonal | |
| Unconjugated | |
| KISS1 | |
| 0610012C01Rik; Eseptin; HH13; KISS1; KiSS-1; KiSS-1 metastasis-suppressor; Kiss1 metastasis-suppressor; Kiss1 variant E1a-E2b-E3; Kiss1 variant E1b-E2a-E3; Kiss1 variant E1b-E2b-E3; kisspeptin; Kisspeptin-1; Kisspeptin-10; Kisspeptin-13; Kisspeptin-14; Kisspeptin-52; Kisspeptin-54; malignant melanoma metastasis-suppressor; metastasis-suppressor KiSS-1; metastatin; metastin; Metastin45-54; MLL septin-like fusion; PP5098; prepro-kisspeptin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3814 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15726 | |
| KISS1 | |
| Recombinant protein corresponding to Human KISS1. Recombinant protein control fragment (Product #RP-95299). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction