missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIRREL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18055-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. The protein encoded by this gene is a synaptic cell adhesion molecule with multiple extracellular immunoglobulin-like domains and a cytoplasmic PDZ domain-binding motif. Mutations in this gene are associated with several neurological and cognitive disorders. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| KIRREL3 | |
| Polyclonal | |
| Unconjugated | |
| KIRREL3 | |
| KIAA1867, Kin of IRRE like protein 3, KIRRE, KIRREL3, MRD4, NEPH2, Nephrin like protein 2, PRO4502 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 315546, 67703, 84623 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BR86, Q8IZU9 | |
| Kirrel3 | |
| KIRREL3 Fusion Protein Ag12573 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction