missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kirrel3/NEPH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | Kirrel3/NEPH2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622906
|
Novus Biologicals
NBP2-38788-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18154699
|
Novus Biologicals
NBP2-38788 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kirrel3/NEPH2 Polyclonal specifically detects Kirrel3/NEPH2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Kirrel3/NEPH2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| KIAA1867MGC126824, kin of IRRE like 3 (Drosophila), Kin of irregular chiasm-like protein 3, kin of IRRE-like protein 3, KIRREPRO4502, MRD4, NEPH2MGC126850, nephrin-like 2, Nephrin-like protein 2 | |
| KIRREL3 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8IZU9 | |
| 84623 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title