missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIRREL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA584870
This item is not returnable.
View return policy
Description
Immunogen sequence: GVPANLTCRS RGDARPTPEL LWFRDGVLLD GATFHQTLLK EGTPGSVEST LTLTPFSHDD GATFVCRARS QALPTGRDTA ITLS
This gene encodes a type I transmembrane protein and member of the immunoglobulin superfamily of cell adhesion molecules. The encoded protein localizes to adherens junctions in pancreatic beta cells and regulates insulin secretion. Autoantibodies against the encoded protein have been detected in serum from patients with type 1 diabetes. This gene may also play a role in glomerular development and decreased expression of this gene has been observed in human glomerular diseases. This gene and the related opposite-strand gene nephrin are regulated by a bidirectional promoter.
Specifications
| KIRREL2 | |
| Polyclonal | |
| Unconjugated | |
| KIRREL2 | |
| DKFZp564A1164, FILTRIN, MGC15718, NEPH3, NLG1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84063 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UWL6 | |
| KIRREL2 | |
| Recombit protein corresponding to Human KIRREL2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction