missing translation for 'onlineSavingsMsg'
Learn More

Kirrel2/NEPH3 Antibody (318311) [DyLight 405], Novus Biologicals™

Product Code. 30734703 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734703 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734703 Supplier Novus Biologicals Supplier No. FAB2564E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kirrel2/NEPH3 Monoclonal antibody specifically detects Kirrel2/NEPH3 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kirrel2/NEPH3
Applications Western Blot
Classification Monoclonal
Clone 318311
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp564A1164, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1kin of irregular chiasm-like 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kirrel2, Gly21-Arg503, Accession # Q6UWL6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84063
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.