missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIRREL2 Monoclonal Antibody (318311)
GREENER_CHOICE

Product Code. 15533732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15533732 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15533732 Supplier Invitrogen™ Supplier No. MA524040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a type I transmembrane protein and member of the immunoglobulin superfamily of cell adhesion molecules. The encoded protein localizes to adherens junctions in pancreatic beta cells and regulates insulin secretion. Autoantibodies against the encoded protein have been detected in serum from patients with type 1 diabetes. This gene may also play a role in glomerular development and decreased expression of this gene has been observed in human glomerular diseases. This gene and the related opposite-strand gene nephrin are regulated by a bidirectional promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIRREL2
Applications Western Blot
Classification Monoclonal
Clone 318311
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene KIRREL2
Gene Accession No. Q6UWL6, Q7TSU7
Gene Alias C330019F22Rik; DKFZp564A1164; FILTRIN; kin of IRRE like 2; kin of IRRE like 2 (Drosophila); kin of irregular chiasm-like 2; kin of irregular chiasm-like protein 2; kin of IRRE-like 2; kin of IRRE-like protein 2; kin of IRRE-like protein 2; LOW QUALITY PROTEIN: kin of IRRE-like protein 2; kirre like nephrin family adhesion molecule 2; KIRREL2; MGC15718; NEPH3; nephrin-like 3; nephrin-like gene 1; nephrin-like protein 3; NLG1; UNQ5827/PRO19646; X kin of IRRE like 2
Gene Symbols KIRREL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kirrel2 Gly21-Arg503.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243911, 84063
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.