missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kirrel1/NEPH1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £366.00
Specifications
| Antigen | Kirrel1/NEPH1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359336
|
Novus Biologicals
NBP3-03509-20UL |
20 μL |
£153.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18372476
|
Novus Biologicals
NBP3-03509-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kirrel1/NEPH1 Polyclonal specifically detects Kirrel1/NEPH1 in Mouse, Rat samples. It is validated for Western Blot.Specifications
| Kirrel1/NEPH1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 55243 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| FLJ10845, kin of IRRE like (Drosophila), MGC129542, MGC129543, NEPH1, nephrin-like protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 17-200 of human Kirrel1/NEPH1 (NP_060710.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title