Learn More
KIRREL Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22232-1-AP-20UL
Description
The KIRREL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIRREL. This antibody recognizes human, mouse, rat antigen. The KIRREL antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| KIRREL | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| kin of IRRE like (Drosophila), Kin of IRRE like protein 1, KIRREL, KIRREL1, NEPH1, Nephrin like protein 1 | |
| 20 μL | |
| 55243 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |