Learn More
KIRREL Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22232-1-AP-150UL
 View more versions of this product
Product Code. 18009675
  • £349.00 / 150µL
Description
The KIRREL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIRREL. This antibody recognizes human, mouse, rat antigen. The KIRREL antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
KIRREL
600 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
kin of IRRE like (Drosophila), Kin of IRRE like protein 1, KIRREL, KIRREL1, NEPH1, Nephrin like protein 1
150 μL
55243
Store at -20°C. Stable for one year after shipment.