missing translation for 'onlineSavingsMsg'
Learn More

Kirenol, TRC

Product Code. 30406895
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406895 1 g -
30389818 250 mg -
30403132 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406895 Supplier Toronto Research Chemicals Supplier No. TRCK5703001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52659-56-0
Molecular Formula C20 H34 O4
Molecular Weight (g/mol) 338.48
Synonym 1,7-Phenanthrenedimethanol, 1,2,3,4,4a,4b,5,6,7,9,10,10a-dodecahydro-3-hydroxy-α7-(hydroxymethyl)-1,4a,7-trimethyl-, (α7R,1R,3S,4aS,4bR,7S,10aS)-, 1,7-Phenanthrenedimethanol, 1,2,3,4,4a,4b,5,6,7,9,10,10a-dodecahydro-3-hydroxy-α7-(hydroxymethyl)-1,4a,7-trimethyl-, [1R-[1α,3β,4aα,4bβ,7α(R*),10aβ]]-, (α7R,1R,3S,4aS,4bR,7S,10aS)-1,2,3,4,4a,4b,5,6,7,9,10,10a-Dodecahydro-3-hydroxy-α7-(hydroxymethyl)-1,4a,7-trimethyl-1,7-phenanthrenedimethanol, Kirenol
IUPAC Name (1R)-1-[(2S,4aR,4bS,6S,8R,8aS)-6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-4,4a,5,6,7,8a,9,10-octahydro-3H-phenanthren-2-yl]ethane-1,2-diol
SMILES C[C@@]1(CO)C[C@@H](O)C[C@@]2(C)[C@@H]3CC[C@@](C)(C=C3CC[C@H]12)[C@@H](O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H34O4/c1-18(17(24)11-21)7-6-15-13(8-18)4-5-16-19(2,12-22)9-14(23)10-20(15,16)3/h8,14-17,21-24H,4-7,9-12H2,1-3H3/t14-,15-,16-,17+,18+,19+,20+/m1/s1
Formula Weight 338.2457
Chemical Name or Material Kirenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.