missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kir7.1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109875
This item is not returnable.
View return policy
Description
Immunogen sequence: IARPKNRAFS IRFTDTAVVA HMDGKPNLIF QVANTRPSPL TSVRVSAVLY QERENGKLYQ TSVDFHLDGI SSDECPFFIF PLTYYHSITP SSPLATLLQH ENPSHFELVV FLSAMQEGTG EICQRRTSYL PSEIMLHHCF ASLLTRGSKG EYQIKMENFD KTVPEFPTPL VSKSPNRTDL DIHINGQSID NFQISETGLT E.
Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. KCNJ13 has a very low single channel conductance, low sensitivity to block by external barium and cesium, and no dependence of its inward rectification properties on the internal blocking particle magnesium.
Specifications
| Kir7.1 | |
| Polyclonal | |
| Unconjugated | |
| KCNJ13 | |
| Inward rectifier K(+) channel Kir7.1; inward rectifier potassium channel 13; Kcnj13; KIR1.4; KIR7.1; LCA16; Potassium channel, inwardly rectifying subfamily J member 13; potassium channel, inwardly rectifying subfamily J, member 13; potassium inwardly-rectifying channel, subfamily J, member 13; potassium voltage-gated channel subfamily J member 13; SVD | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100040591, 3769 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.56 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O60928, P86046 | |
| KCNJ13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-360 of human KCNJ13 (NP_002233.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction