missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir6.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16920-1-AP-150UL
This item is not returnable.
View return policy
Description
This receptor is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by extracellular barium By similarity. Subunit of ATP-sensitive potassium channels (KATP). Can form cardiac and smooth muscle-type KATP channels with ABCC9. KCNJ11 forms the channel pore while ABCC9 is required for activation and regulation.Specifications
| Kir6.2 | |
| Polyclonal | |
| Unconjugated | |
| KCNJ11 | |
| BIR, HHF2, IKATP, KCNJ11, KIR6.2, PHHI, TNDM3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16514, 3767, 83535 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P70673, Q14654, Q61743 | |
| Kcnj11 | |
| Kir6.2 Fusion Protein Ag10262 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction