missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR5.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | KIR5.1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
KIR5.1 Polyclonal specifically detects KIR5.1 in Human samples. It is validated for Western Blot.Specifications
| KIR5.1 | |
| Polyclonal | |
| Rabbit | |
| NP_061128 | |
| 3773 | |
| Synthetic peptide directed towards the middle region of human KCNJ16. Peptide sequence RESCTSDTKARRRSFSAVAIVSSCENPEETTTSATHEYRETPYQKALLTL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BIR9, Inward rectifier K(+) channel Kir5.1, inward rectifier K+ channel KIR5.1, inward rectifier potassium channel 16, KIR5.1, MGC33717, Potassium channel, inwardly rectifying subfamily J member 16, potassium inwardly-rectifying channel, subfamily J, member 16 | |
| KCNJ16 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title