missing translation for 'onlineSavingsMsg'
Learn More

KIR3DX1 Antibody (2H4), Novus Biologicals™

Product Code. 18408309 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408309 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408309 Supplier Novus Biologicals Supplier No. H00090011M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIR3DX1 Monoclonal antibody specifically detects KIR3DX1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR3DX1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2H4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias FLJ00060, killer cell immunoglobulin-like receptor, three domains, X1, KIR3DL0, LENG12
Host Species Mouse
Immunogen KIR3DX1 (AAH33195.1, 1 a.a. ∽ 78 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MANTEPTEGQRTDEEEPAAEETQEIIYAQLNHQALSQTGFPPASQCPHYLSEDPSIYITVHQAQAEARAAPSLWHKGH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90011
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.