Learn More
Invitrogen™ KIR3DS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597295
Description
Immunogen sequence: FNMSPVTTAH AGNYTCRGSH PHSPTGWSAP SNPMVIMVTG NHRKPSLLAH PGPLVKSGER VILQCWSDIM FEHFFLHKEW ISKDPSRLVG QIHDGVSKAN FSIGSMMRAL AGTYRCYGSV THTPYQLSAP SDPLDIVVTG LYEKPSLSAQ PGPKVQAGES VTLSCSSRSS YDMYHLSREG GAHERRLPAV RKVNRTFQAD FPLGPATHGG TYRCFGSFRH SPYEWSDPSD PLLVSVTGNP SSSWPSPTEP SSKSGNLRHL H; Positive Samples: Jurkat, A-549, THP-1, BT-474, B-cell, Mouse liver, Mouse kidney, Mouse pancreas, Rat liver; Cellular Location: Cell membrane, Single-pass type I membrane protein.
The killer immunoglobulin-like receptors (KIRs) on natural killer (NK) cells regulate the inhibition and activation of NK-cell responses through recognition of human leukocyte antigen (HLA) class I molecules. KIR3DL1, a receptor for HLA-B antigens with the Bw4 allele, transmits an inhibitory signal to prevent killer cell-mediated cytoxicity. KIR3DL1 encodes a 444 amino acid type I transmembrane protein, containing three immunoglobulin-like C2-type domains.
Specifications
| KIR3DS1 | |
| Polyclonal | |
| Unconjugated | |
| KIR3DS1 | |
| CD158E2; killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin-like receptor 3DS1; killer cell immunoglobulin-like receptor KIR3DS1; killer cell immunoglobulin-like receptor, three domains, short cytoplasmic tail, 1; killer Ig receptor; KIR-123FM; KIR3DS1; KIR-G1; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 10; NKAT10; NKAT-10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3813 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.97 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14943 | |
| KIR3DS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-340 of human KIR3DS1 (NP_0010770081). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.