missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIR3DS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576457
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The killer immunoglobulin-like receptors (KIRs) on natural killer (NK) cells regulate the inhibition and activation of NK-cell responses through recognition of human leukocyte antigen (HLA) class I molecules. KIR3DL1, a receptor for HLA-B antigens with the Bw4 allele, transmits an inhibitory signal to prevent killer cell-mediated cytoxicity. KIR3DL1 encodes a 444 amino acid type I transmembrane protein, containing three immunoglobulin-like C2-type domains.
Specifications
| KIR3DS1 | |
| Polyclonal | |
| Unconjugated | |
| KIR3DS1 | |
| CD158E2; killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin-like receptor 3DS1; killer cell immunoglobulin-like receptor KIR3DS1; killer cell immunoglobulin-like receptor, three domains, short cytoplasmic tail, 1; killer Ig receptor; KIR-123FM; KIR3DS1; KIR-G1; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 10; NKAT10; NKAT-10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3813 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14943 | |
| KIR3DS1 | |
| Recombinant fusion protein of human KIR3DS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction