Learn More
Invitrogen™ KIR3DL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587606
Description
Immunogen sequence: QDKPFLSAWP GTVVSEGQHV TLQCRSRLGF NEFSLSKEDG MPVPELYNRI FRNSFLMGPV TPAHAGTYRC CSSHPHSPTG WSAPSNPVVI MVTGVHRKPS LLAHPGPLVK SGETVILQCW SDVRFERFLL HREGITEDPL RLVGQLHDAG SQVNYSMGPM TPALAGTYRC FGSVTHLPYE LSAPSDPLDI VVVGLYGKPS LSAQPGPTVQ AGENVTLSCS SRSLFDIYHL SREAEAGELR LTAVLRVNGT FQANFPLGPV THGGNYRCFG SFRALPHAWS DPSDPLPVSV TGNSRNL; Positive Samples: HepG2, SGC-7901; Cellular Location: Cell membrane, Single-pass type I membrane protein.
KIR3DL3 is transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. This protein are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC).
Specifications
| KIR3DL3 | |
| Polyclonal | |
| Unconjugated | |
| KIR3DL3 | |
| CD158 antigen-like family member Z; CD158Z; killer cell Ig-like receptor KIR3DL7; killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3; killer cell immunoglobulin-like receptor 3DL3; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 3; killer cell inhibitory receptor 1; killer-cell immunoglobulin-like receptor 3DL3; KIR2DS2; KIR3DL3; KIR3DL7; KIR44; KIRC1; natural killer cell receptor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 115653 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N743 | |
| KIR3DL3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-322 of human KIR3DL3 (NP_7031443). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.