missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DL3/CD158z Antibody (CH21), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21499R
This item is not returnable.
View return policy
Description
KIR3DL3/CD158z Monoclonal antibody specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry
Specifications
| KIR3DL3/CD158z | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 115653 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| CH21 | |
| Flow Cytometry | |
| CD158Z;Killer cell immunoglobulin-like receptor 3DL3;KIR2DL5B;KIR3DL7;KIR44;KIRC1 | |
| Human KIR3DL3/CD158z transfectants | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction