missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DL3/CD158z Antibody (1136B), PE/Cy7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8919PECY7
This item is not returnable.
View return policy
Description
KIR3DL3/CD158z Monoclonal antibody specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry
Specifications
| KIR3DL3/CD158z | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 115653 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry | |
| 1136B | |
| Flow Cytometry | |
| CD158Z;Killer cell immunoglobulin-like receptor 3DL3;KIR2DL5B;KIR3DL7;KIR44;KIRC1 | |
| Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z, Met1-Leu322, Accession # Q8N743 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction