missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DL2/CD158k Antibody (lacutamab) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28581JF525
This item is not returnable.
View return policy
Description
KIR3DL2/CD158k Monoclonal antibody specifically detects KIR3DL2/CD158k in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| KIR3DL2/CD158k | |
| Monoclonal | |
| Janelia Fluor 525 | |
| killer cell immunoglobulin-like receptor 3DL2, CD158K, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2, NKAT4, NKAT-4, NKAT4B, p140 | |
| KIR3DL2/ CD158k | |
| 0.1 mL | |
| Immunology | |
| 3812 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| lacutamab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction