missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KIR3DL1 Monoclonal antibody specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry, CyTOF, Neutralization
Specifications
Specifications
| Applications | Flow Cytometry, CyTOF |
| Conjugate | HRP |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?