missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DL1 Antibody (MM0443-1L34), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11763F
This item is not returnable.
View return policy
Description
KIR3DL1 Monoclonal specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR3DL1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| KIR3DL1 | |
| Human recombinant KIR3DL1 | |
| 0.1 mL | |
| Primary | |
| This antibody was selected for its ability to specifically detect human KIR3DL1. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| MM0443-1L34 | |
| Flow Cytometry | |
| AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, NKAT-3, NKAT3KIR, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 | |
| Mouse | |
| Protein G purified | |
| Immunology | |
| 3811 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction