missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir3.4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03006-20UL
This item is not returnable.
View return policy
Description
Kir3.4 Polyclonal specifically detects Kir3.4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Kir3.4 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3762 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Cardiac inward rectifier, CIRKIR3.4, G protein-activated inward rectifier potassium channel 4, GIRK-4, GIRK4Kir3.4, Heart KATP channel, Inward rectifier K(+) channel Kir3.4, inward rectifier K+ channel KIR3.4, IRK-4, KATP-1, KATP1cardiac ATP-sensitive potassium channel, LQT13, Potassium channel, inwardly rectifying subfamily J member 5, potassium inwardly-rectifying channel, subfamily J, member 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Kir3.4 (NP_000881.3). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction