missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kir3.3 (KCNJ9) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101594
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCNJ9.
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins. It associates with another G-protein-activated potassium channel to form a heteromultimeric pore-forming complex. [provided by RefSeq, Jul 2008].
Specifications
| Kir3.3 (KCNJ9) | |
| Polyclonal | |
| Unconjugated | |
| KCNJ9 | |
| 1700085N21Rik; G protein-activated inward rectifier potassium channel 3; G protein-coupled inward rectifier potassium channel; Girk3; GIRK-3; Inward rectifier K(+) channel Kir3.3; inwardly rectifier K(+) channel Kir3.3; inwardly rectifier K+ channel KIR3.3; KCNJ9; Kir3.3; mbGIRK3; Potassium channel, inwardly rectifying subfamily J member 9; potassium channel, inwardly rectifying subfamily J, member 9; potassium inwardly rectifying channel subfamily J member 9; potassium inwardly-rectifying channel subfamily J9; potassium inwardly-rectifying channel, subfamily J, member 9; potassium voltage-gated channel subfamily J member 9; si:dkey-42d18.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116560, 16524, 3765 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P48543, Q63511, Q92806 | |
| KCNJ9 | |
| A synthesized peptide derived from human KCNJ9(Accession Q92806), corresponding to amino acid residues E324-A374. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction