missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir3.3 (KCNJ9) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577599
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins. It associates with another G-protein-activated potassium channel to form a heteromultimeric pore-forming complex. [provided by RefSeq, Jul 2008].
Specifications
| Kir3.3 (KCNJ9) | |
| Polyclonal | |
| Unconjugated | |
| KCNJ9 | |
| GIRK3, Kcnj9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116560, 16524, 3765 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.75 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P48543, Q63511, Q92806 | |
| Kcnj9 | |
| Peptide (C)RLDAHLYWSIPSRLDEKV, corresponding to residues 344-361 of rat Kir 3.3 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction