Learn More
Kir3.2 (KCNJ6) Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Brand: Invitrogen PA5111799
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). GST fusion protein sequence is: ELANRAEVPL SWSVS SKLN QHAELETEEE EKNPEELTER NG Homology: Rat - 40/41, human - 37/41 amino acid residues identical.
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins and may be involved in the regulation of insulin secretion by glucose. It associates with two other G-protein-activated potassium channels to form a heteromultimeric pore-forming complex.
Specifications
| Kir3.2 (KCNJ6) | |
| Polyclonal | |
| Unconjugated | |
| KCNJ6 | |
| BIR1; G protein-activated inward rectifier potassium channel 2; GIRK2; GIRK-2; hiGIRK2; Inward rectifier K(+) channel Kir3.2; KATP2; KATP-2; KCNJ6; Kcnj7; KIR3.2; KPLBS; MGC126596; Potassium channel, inwardly rectifying subfamily J member 6; W; weaver; wv | |
| Guinea Pig | |
| Antigen affinity chromatography | |
| RUO | |
| 16522, 25743 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P48542, P48550 | |
| KCNJ6 | |
| GST fusion protein corresponding to residues 374-414 of mouse Kir3.2 (GIRK2) | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.