missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir3.1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | Kir3.1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18327884
|
Novus Biologicals
NBP3-03002-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362476
|
Novus Biologicals
NBP3-03002-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kir3.1 Polyclonal specifically detects Kir3.1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Kir3.1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 3760 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| G protein-activated inward rectifier potassium channel 1, GIRK-1, GIRK1Kir3.1, Inward rectifier K(+) channel Kir3.1, inward rectifier K+ channel KIR3.1, KGA, KIR3.1, Potassium channel, inwardly rectifying subfamily J member 3, potassium inwardly-rectifying channel, subfamily J, member 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Kir3.1 (NP_002230.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title