missing translation for 'onlineSavingsMsg'
Learn More

Kir3.1 Antibody (OTI1G3), DyLight 594, Novus Biologicals™

Product Code. 18428062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428062 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428062 Supplier Novus Biologicals Supplier No. NBP271065DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kir3.1 Monoclonal antibody specifically detects Kir3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir3.1
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1G3
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias G protein-activated inward rectifier potassium channel 1, GIRK-1, GIRK1Kir3.1, Inward rectifier K(+) channel Kir3.1, inward rectifier K+ channel KIR3.1, KGA, KIR3.1, Potassium channel, inwardly rectifying subfamily J member 3, potassium inwardly-rectifying channel, subfamily J, member 3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 279-501 of human Kir3.1 (NP_002230) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3760
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.