missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir3.1 Antibody (OTI1G3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71065AF350
This item is not returnable.
View return policy
Description
Kir3.1 Monoclonal antibody specifically detects Kir3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Kir3.1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| G protein-activated inward rectifier potassium channel 1, GIRK-1, GIRK1Kir3.1, Inward rectifier K(+) channel Kir3.1, inward rectifier K+ channel KIR3.1, KGA, KIR3.1, Potassium channel, inwardly rectifying subfamily J member 3, potassium inwardly-rectifying channel, subfamily J, member 3 | |
| Recombinant protein fragment corresponding to amino acids 279-501 of human Kir3.1 (NP_002230) produced in E.coli. | |
| 0.1 mL | |
| Cardiovascular Biology, Neuroscience | |
| 3760 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1G3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction