missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DS5/CD158g Antibody (1165A) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8920JF585
This item is not returnable.
View return policy
Description
KIR2DS5/CD158g Monoclonal antibody specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry
Specifications
| KIR2DS5/CD158g | |
| Monoclonal | |
| Janelia Fluor 585 | |
| killer cell immunoglobulin-like receptor 2DS5, CD158G, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9 | |
| NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g, His22-His245, Accession # Q14953 | |
| RUO | |
| 3810 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 1165A | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction