missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DS2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82102PCP
This item is not returnable.
View return policy
Description
KIR2DS2 Polyclonal antibody specifically detects KIR2DS2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| KIR2DS2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| 183ActI, CD158b, CD158J, cl-49, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2, NKAT5, p58 KIR | |
| KIR2DS2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human KIR2DS2. The immunogen is located within the last 50 amino acids of KIR2DS2. Amino Acid Squence: EPAGNRTVNSEDSDEQDH | |
| 0.1 mL | |
| Immunology | |
| 100132285 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction