missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15005
This item is not returnable.
View return policy
Description
KIR2DS2 Polyclonal specifically detects KIR2DS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| KIR2DS2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| 183ActI, CD158b, CD158J, cl-49, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2, NKAT5, p58 KIR | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 100132285 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P43631 | |
| KIR2DS2 | |
| Recombinant protein encompassing a sequence within the center region of human KIR2DS2. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction