missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL5/CD158f Antibody (UP-R1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62222PCP
This item is not returnable.
View return policy
Description
KIR2DL5/CD158f Monoclonal specifically detects KIR2DL5/CD158f in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR2DL5/CD158f | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| KIR2DL5A | |
| Human CD158f-Ig fusion protein | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-59 recognizes neuraminidase-sensitive epitope on CD43 (Leukosialin), a 95-135 kDa type I transmembrane glycoprotein (mucin-type) which is involved in lymphocyte activation. CD43 is expressed by platelets and at high levels on the surface of all leukocytes; it is negative on resting B lymphocytes and erythrocytes. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| UP-R1 | |
| Flow Cytometry | |
| CD158F, CD158F1, killer cell immunoglobulin-like receptor 2DL5A, killer cell immunoglobulin-like receptor KIR2DL5A, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A, killer-cell Ig-like receptor, KIR2DL5, KIR2DL5.1, KIR2DL5.3, KIR2DL5B | |
| Mouse | |
| Protein A purified | |
| Immunology | |
| 57292 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction