missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL5/CD158f Antibody (UP-R1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62222
This item is not returnable.
View return policy
Description
KIR2DL5/CD158f Monoclonal antibody specifically detects KIR2DL5/CD158f in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| KIR2DL5/CD158f | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| UP-R1 | |
| Flow Cytometry 1-4 μg/mL, Immunocytochemistry | |
| CD158F, CD158F1, killer cell immunoglobulin-like receptor 2DL5A, killer cell immunoglobulin-like receptor KIR2DL5A, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A, killer-cell Ig-like receptor, KIR2DL5, KIR2DL5.1, KIR2DL5.3, KIR2DL5B | |
| Human CD158f-Ig fusion protein | |
| 0.1 mg | |
| Immunology | |
| 57292 | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction