missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL4/CD158d Antibody (mAb33), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-50052AF750
This item is not returnable.
View return policy
Description
KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| KIR2DL4/CD158d | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| KIR2DL4 | |
| NK3.3 cells and KIR2DL4-Ig fusion protein | |
| 0.1 mL | |
| Primary | |
| The mouse monoclonal antibody mAb#33 (also known as mAb 33 or 33) recognizes extracellular portion of CD158d/ KIR2DL4, a 45 kDa NK cell marker. Cell surface expression and function of CD158d/ KIR2DL4 depends on genotype of particular individuals. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| mAb33 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor | |
| Mouse | |
| Protein A purified | |
| Immunology | |
| 3805.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction