missing translation for 'onlineSavingsMsg'
Learn More

KIR2DL4/CD158d Antibody (33) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™

Product Code. 18345213 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345213 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345213 Supplier Novus Biologicals Supplier No. NBP305290

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL4/CD158d
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, Immunocytochemistry
Classification Monoclonal
Clone 33
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 2 to 6 μg/mL, Immunoprecipitation, Functional, Immunocytochemistry
Formulation Low endotoxin azide free phosphate buffered saline (PBS) solution, 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test.
Gene Alias 103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor
Host Species Mouse
Immunogen NK3.3 cells and KIR2DL4-Ig fusion protein
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3805
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.