missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL4/CD158d Antibody (2H6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04312F
This item is not returnable.
View return policy
Description
KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for ELISA.
Specifications
| KIR2DL4/CD158d | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| KIR2DL4 | |
| Recombinant human KIR2DL4 (44-202 aa) purified from E. coli | |
| 0.1 mL | |
| Immunology | |
| 3805 | |
| Human | |
| Purified |
| ELISA | |
| 2H6 | |
| ELISA | |
| 103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Anti-human KIR2DL4 mAb, clone 2H6, is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c immunized with recombinant human KIR2DL4 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction