missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL4/CD158d Antibody (181703) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2238JF669
This item is not returnable.
View return policy
Description
KIR2DL4/CD158d Monoclonal antibody specifically detects KIR2DL4/CD158d in Human samples. It is validated for Flow Cytometry,Agonist Activity,CyTOF-reported
Specifications
| KIR2DL4/CD158d | |
| Monoclonal | |
| Janelia Fluor 669 | |
| 103AS, 15.212, CD158 antigen-like family member D, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL4/CD158d, Accession # Q99706 | |
| 0.5 mL | |
| Immunology | |
| 3805 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Agonist Activity, CyTOF | |
| 181703 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction