missing translation for 'onlineSavingsMsg'
Learn More

KIR2DL3/CD158b2 Antibody (MM0440-8P9) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£153.00 - £427.00

Specifications

Antigen KIR2DL3/CD158b2
Clone MM0440-8P9
Concentration LYOPH
Applications CyTOF, Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18050867
View Documents
Novus Biologicals
NBP2-11760-0.025MG
0.025 mg
£153.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18715023
View Documents
Novus Biologicals
NBP2-11760
£427.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KIR2DL3/CD158b2 Monoclonal specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

KIR2DL3/CD158b2
LYOPH
Monoclonal
Purified
RUO
Human
CD158 antigen-like family member B2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, NKAT2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
KIR2DL3
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
MM0440-8P9
CyTOF, Flow Cytometry
Unconjugated
Mouse
Apoptosis
ACI49719.1
3804
IgG2
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.