missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL3/CD158b2 Antibody (180701) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2014JF669
This item is not returnable.
View return policy
Description
KIR2DL3/CD158b2 Monoclonal antibody specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for Flow Cytometry,CyTOF-reported
Specifications
| KIR2DL3/CD158b2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD158 antigen-like family member B2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, NKAT2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2, Accession # P43628 | |
| 0.5 mL | |
| Apoptosis | |
| 3804 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 180701 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction