missing translation for 'onlineSavingsMsg'
Learn More

KIR2DL3/CD158b2 Antibody (014), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18339440 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339440 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339440 Supplier Novus Biologicals Supplier No. NBP290149AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KIR2DL3/CD158b2 Monoclonal specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL3/CD158b2
Applications ELISA
Classification Monoclonal
Clone 14
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD158 antigen-like family member B2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, NKAT2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human KIR2DL3/CD158b2 (AAX23102.1; Met1-His245).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 3804
Test Specificity Has cross-reactivity in ELISA with: Human KIR2DL4 No cross-reactivity in ELISA with: Human KIR2DL1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.