missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIR2DL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575623
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several framework genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
Specifications
| KIR2DL2 | |
| Polyclonal | |
| Unconjugated | |
| KIR2DL2 | |
| CD158 antigen-like family member B1; CD158b; CD158B1; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 2; killer cell immunoglobulin-like receptor 2DL2; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 2; KIR2DL2; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 6; NKAT6; NKAT-6; p58 natural killer cell receptor clone CL-43; p58 NK receptor CL-43; p58.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3803 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43627 | |
| KIR2DL2 | |
| Synthetic peptide corresponding to residues in Human KIR2DL2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction