missing translation for 'onlineSavingsMsg'
Learn More

KIR2DL2 Polyclonal Antibody

Product Code. 16690355 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16690355 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16690355 Supplier Thermo Scientific Supplier No. PA5103908
  • ONLINE EXCLUSIVE PRICE
    £449.00
    £354.71 / 100µL
    Save £94.29
    21% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KIR2DL2 Polyclonal Antibody

KIR2DL2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIR2DL2
Gene Accession No. P43627
Gene Alias CD158 antigen-like family member B1; CD158b; CD158B1; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 2; killer cell immunoglobulin-like receptor 2DL2; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 2; KIR2DL2; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 6; NKAT6; NKAT-6; p58 natural killer cell receptor clone CL-43; p58 NK receptor CL-43; p58.2
Gene Symbols KIR2DL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIR2DL2(Accession P43627), corresponding to amino acid residues T146-Y196.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3803
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.